Savant™ SpeedVac™ Intergrated Vacuum Concentrator, 220 V
Supplier: Thermo Fisher Scientific
SPD2030P1-220 SPD2030A-220
76210-544EA 39816 CAD
76210-544 76210-552
Savant™ SpeedVac™ Intergrated Vacuum Concentrator, 220 V
Evaporators Centrifugal Evaporators
The Savant SpeedVac SPD1030 system is designed for medium capacity (120×1.5 ml tubes and other formats) and the SpeedVac SPD2030 system is ideal for higher capacity (200×1.5 ml tubes and other formats).
  • 220 V model
  • Fully integrated
  • Medium capacity
  • Variety of rotors
  • Radiant glass cover
  • PTFE coated aluminum chamber
A variety of rotors are available to support sample formats from micro-centrifuge tubes, vials and microplates to large-volume flasks. The unit will fit in a standard laboratory fume hood.
Caution: 220 V model. Corresponding model with voltage suitable for use in US (76210-540 and 76210-548) are also available, please see separate product page.