Bovine Serum Albumin (BSA)
Supplier: MilliporeSigma
2910-25GM
2930-100GM
2960-500GM
CA-EM2910EA
1382.57
CAD
CA-EM2910
CA-EM2930
CA-EM2960
CA-EM2980
Bovine Serum Albumin (BSA)
Proteins and Peptides
Fraction V, heat shock isolation.
- DNase-, RNase-, and protease-free