VWR® Tweezers, PTFE
Supplier: VWR International
89026-418 89026-420 89026-424 89026-426
89026-418EA 164.26 CAD
89026-418 89026-420 89026-424 89026-426
VWR® Tweezers, PTFE
Forceps
Tweezers have a smooth PTFE non-stick surface.
  • PTFE construction
  • Non-contaminating
  • Inert
For use in temperatures of 0 to 280°C (32 to 540°F). Non-contaminating and inert to virtually all chemical reagents.