Agarose, OmniPur®, Millipore®

Supplier: MilliporeSigma

2120-100GM 2125-500GM
CA-EM2120EA 1557.89 CAD
CA-EM2120 CA-EM2125
Agarose, OmniPur®, Millipore®
Agarose

Suitable for a wide range of nucleic acid and protein gel applications. Of average gel strength and standard melting and gelling ranges. Low EEO for shorter electrophoretic runs. Molecular biology grade.


  • Molecular Biology Grade


Specifications:
DNAse: none detected
RNAse: none detected
Protease: none detected
Sulfate (SO4): ≤0.15%
EEO (-mr): ≤0.1
Gelling range (1%): 36-39°C
Gel strength (1%): 1200 g/cm2 min
Melting point (1%): 87-89°C


Caution: Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR