Acetone ≥99.9% (dried basis), Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

PP010-19 PP010-204
CABJPP010-19EA 752.93 CAD
CABJPP010-19 CABJPP010-204
Acetone ≥99.9% (dried basis), Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Acetone

For specialty organic synthesis, preparative scale HPLC, glassware cleaning, and applications requiring ACS reagent grade solvent. The 54gal. drum is individually analyzed with certificate attached. Soluble in water.


  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Aldehyde (as HCHO) 0.002 % max.
Water by Karl Fischer Titration 0.5 % max.
Assay by GC (excluding water) 99.9 % min.
Isopropyl Alcohol 0.05 % max.
Methanol 0.05 % max.
Refractive Index (20ºC) 1.3576 - 1.3596
Residue after Evaporation 5 mg/L max.
Substances Reducing Permanganate Passes test
Titrable Acid 0.3 µeq/g max.
Titrable Base 0.6 µeq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR