Acetonitrile ≥99.5% ACS analytical reagent, general laboratory use

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

LP014TC19 LP014-4 LP014-204 LP014-4P
CABJLP014TC19EA 2556.33 CAD
CABJLP014TC19 CABJLP014-4 BJLP014-204 BJLP014-4P
Acetonitrile ≥99.5% ACS analytical reagent, general laboratory use
Acetonitrile

ACS general use certified, reagent grade


  • Low residue
  • Low water
  • High GC purity

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.005 % max.
Tirable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
Water by Karl Fischer Titration 0.3 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR