Acetone ≥99.5% ACS analytical reagent, general laboratory use

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

LP010-19 LP010-4 LP010-19 LP010-204 LP010-4P
CABJLP010D19EA 903.78 CAD
CABJLP010D19 CABJLP010-4 BJLP010-19 BJLP010-204 BJLP010-4P
Acetone ≥99.5% ACS analytical reagent, general laboratory use
Acetone

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56 °C
Melting Pt: –94.8 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Aldehyde (as HCHO) 0.002 % max.
Water by Karl Fischer Titration 0.5 % max.
Color (APHA) 10 max.
Isopropyl Alcohol 0.05 % max.
Methanol 0.05 % max.
Residue after Evaporation 0.001 % max.
Solubility in Water Passes test
Substances Reducing Permanganate Passes test
Titrable Acid 0.3 µeq/g max.
Titrable Base 0.6 µeq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR