Heat-Tolerant/Cleanroom Applicator Brushes, Gordon Brush
Supplier: Gordon Brush
250NADG-12 375NADG-12 499NADG-12
22940-590EA 106.86 CAD
22940-590 22940-592 22940-588
Heat-Tolerant/Cleanroom Applicator Brushes, Gordon Brush
Brushes
Pencil shaped applicator brushes with rigid acetal handle and Nylon bristles.
  • Can withstand most sterilizing environments
  • Bristles are epoxy set with high temperature adhesive that is autoclavable
Brushes must be cleaned prior to cleanroom use.
The brush is typically used in situations where a round brush is needed to clean fine areas, and for applying liquids or fluids in a high temperature setting.