Nalgene® Clearboy™ Carboys, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
2251-0020 2251-0050 2317-0020 2317-0050
16101-164CS 1179.11 CAD
16101-164 16101-186 16101-608 16101-641
Nalgene® Clearboy™ Carboys, Polycarbonate, Thermo Scientific
Bottles Carboys
Useful for media preparation and culture, especially when light is required.
  • Leakproof
  • Autoclavable
For replacement caps, see cap 16225-234 in 16219-122 series.
Note: Autoclave using a properly vented closure. For carboys with spigots, autoclave only when empty.
Ideal for storing or dispensing reagents or distilled water. Practical for use in cold rooms, even at subfreezing temperatures. Carboys are graduated in 1 gal. and 5 L increments. Carboys without spigot are graduated to contain; carboys with spigot are graduated to deliver. Cap included. Cap size: 83B.