Peroxigard™ Cleaners & Disinfectants, Lighthouse
Supplier: Unimed-Midwest
PRTU242101 PRTU242105 PRTU242111 PWIP242221
77441-328CS 165.99 CAD
77441-328 77441-330 77441-332 77441-334
Peroxigard™ Cleaners & Disinfectants, Lighthouse
Cleaning Agents Detergents and Surface Cleansers
Peroxigard™ Ready-To-Use eliminates variables in your cleaning and disinfection protocols, driving efficiency by removing the risks associated with dilution, activation and/or deactivation. Ideal for cleaning and disinfecting bio-safety cabinets, laboratory equipment and tools, cages, work stations, animal housing rooms, high-touch surfaces, and surgical suites.
  • Efficacy - Cleans and achieves disinfection contact time in 1 minute* against pathogens of concern in a single application.
  • Safety - Eliminates the need to interact with hazardous concentrates. Designed with animal safety in mind. Fragrance free, non-irritating, non-corrosive and superior metal compatibility.
  • Simplicity - Removes the risks associated with diluting concentrates and eliminates the complexity of using multiple products at various dilution ratios
Peroxigard™ Ready to Use Liquid and Wipes utilize Accelerated Hydrogen Peroxide®, a patented and synergistic blend of commonly used ingredients, that when combined with low levels of hydrogen peroxide create an ideal one-step cleaner and disinfectant. Peroxigard™ RTU and Wipes disinfect in only 1 minute*, removing the risk of infection associated with improperly diluting and not achieving contact time compliance. The wipe product is a convenient way to remove variables by providing a consistent amount of liquid in every wipe.
For Use In:
• Vivariums
• Breeding facilities
• Lab animal research facilities
• Laboratories
For Use On:
• Biosafety cabinets
• Lab benches and exam tables
• Diagnostic and laboratory equipment
• Cages, crates, and carts
Available format:
• Ready-To-Use Liquid (EPA Reg No. 74559-9) 32 oz, 1 gal, 5 gal, 30 gal, 55 gal
• Presaturated Wipes (EPA Reg No. 74559-10) 160 ct. 6x7
Caution: Shelf life 2 years (Opened/Unopened)