Savant™ SpeedVac™ Modular Vacuum Concentrators, 230 V
Supplier: Thermo Fisher Scientific
SPD210-230 SPD210P1-230 SPD210P2-230
76210-402EA 13872.4 CAD
76210-402 76210-566 76210-570
Savant™ SpeedVac™ Modular Vacuum Concentrators, 230 V
Evaporators Centrifugal Evaporators
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
  • 230 V model
  • Kit
  • Medium capacity
  • Variety of rotors
  • Glass safety cover
  • PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
More Product Information