Ammonia solution 28 - 30%, ARISTAR® ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Ammonia aqueousAmmonium hydroxide solutionAmmonia waterAmmonium hydroxide

048500-80603 048500-80602 048500-80617 048500-80601 BDH3126-2.2LP 048500-80401 3022 048500-24401 3018 3016 3020 3014
CABDH3126-2.2PCS 860.92 CAD
BDH3014-500MLP BDH3020-3.8LP CABDH3126-2.2P BDH3022-194L BDH3016-2.5LG BDH3126-2.2LP CABDH30162.5LG BDH3018-2.5LPC CABDH3018PC CABDH3014P CABDH3022194L CABDH30203.8LP
Ammonia solution 28 - 30%, ARISTAR® ACS, VWR Chemicals BDH®
Ammonia solution

Clear and free from particulates.

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless, free from susp. matter
Assay (NH3) 28.0 - 30.0%
Carbon dioxide (CO2) 0.002% max.
Chloride (Cl) 0.5 ppm max.
Heavy Metals (as Pb) 0.5 ppm max.
Iron (Fe) 0.2 ppm max.
Nitrate (NO3) 2 ppm max.
Phosphate (PO4) 2 ppm max.
Residue After Ignition 0.002% max.
Substances Reducing Permangana To pass test
Sulfate (SO4) 2 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR