Acid Resistant Refrigerated CentriVap Centrifugal Vacuum Concentrators, 230 V
Supplier: Labconco
7310032 7310034 7310039 7310042
89187-286EA 26828.4 CAD
89187-286 89187-288 89187-290 89187-292
Acid Resistant Refrigerated CentriVap Centrifugal Vacuum Concentrators, 230 V
Evaporators Centrifugal Evaporators
The acid-resistant refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature.
  • 230 V model
This exclusive concentrator combines a refrigeration system that cools to –4 °C with a heater that warms to 100 °C to achieve the users desired set point temperatures. It also features a corrosion-resistant, Teflon-coated chamber and Hastelloy shaft to protect against samples containing corrosive solvents.