Acetonitrile ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH1103-19L
BDH1103-4LG
BDH1103-4LP
BDH1103-19LEA
1411.1
CAD
BDH1103-19L
BDH1103-4LG
BDH1103-4LP
Acetonitrile ≥99.5% ACS, VWR Chemicals BDH®
Acetonitrile
Clear liquid.
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Appearance | Clear |
Water (K.F.) | Less than 0.30% |
Residue | Less than 0.005% (0.0010g/50mL) |
GC Purity | Greater than 99.5% Less than 10 |
Color (APHA) | Less than 10 |
Titrable acid | Less than 8 meq/g (No more than 8.0 mL of 0.01N sodium hydroxide) |
Titrable base | Less than 0.6 meq/g (No more than 4.5 mL of 0.01N hydrochloric acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...