Orbital Shakers, 230 V, KS 260
Supplier: IKA Works
2980200 2980300
33998-358EA 7340.7 CAD
33998-358 33998-362
Orbital Shakers, 230 V, KS 260
Shakers Orbital Shakers
For a variety of applications including medical diagnostics, hematology, water and wastewater testing, and dairy farming
  • 230 V model
Basic model features electronic adjustment of speed, and an LED line speed display. Control model features a digital display for speed, timer function, and operating modes. All functions of the Control model can be controlled and documented with labworldsoft* software via an integral RS-232 interface.
The universal platform holds various types of vessels and features variable clamping rolls that allow for universal adaptation. Universal platform surface dimensions: 32×32 cm (12⁹/₁₆×12⁹/₁₆"). The flask clamp platform is for shaking flasks, Erlenmeyer flasks, and bottles with a round cross-section. Accommodates fifty-six 25 ml flasks, twenty-three 50 or 100 ml flasks, eleven 200 or 250 ml flasks, or nine 500 ml flasks. The dish attachment is for smooth movement of cell cultures, nutrient media, petri dishes, culture bottles, and vessels with a low center of gravity. Dish platform surface dimensions: 32×32 cm (12⁹/₁₆×12⁹/₁₆").
May be used inside refrigerators or incubators within the specified temperature range. Features include a 50 W motor, 10 mm (²/₈") orbit diameter, and an electronic time switching clock. Clock can be set for timed or continuous operation.
Ordering information: Control models include labworldsoft software, a 2.5 m (8.2') PC 1.5 cable, and a PC 2.2 adapter. Universal platform includes four clamping rolls and eight fastening screws.
Caution: 230 V models. Corresponding models with voltage suitable for use in US (733998-360, 33998-364) are also available, please see separate product page.