Antimony ≥99.9999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.1 in)

Supplier: Thermo Scientific Chemicals
Warning

11071-09 11071-18 11071-30
CAAA11071-09EA 126.24 CAD
CAAA11071-09 CAAA11071-18 CAAA11071-30
Antimony ≥99.9999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.1 in)
Antimony

MDL: MFCD00134030


Purity: 99.9999% CAS No.: 7440-36-0 Odor: Odorless MDL No.: MFCD00134030

Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733

Order Now

Specification Test Results

Assay (by Metals Basis) 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR