Aluminium 99,999 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

10747-G5 10747-G9 10747-H4 10747-H8
CAAA10747-G5EA 114.71 CAD
CAAA10747-G5 CAAA10747-G9 CAAA10747-H4 CAAA10747-H8
Aluminium 99,999 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈2.1g/m, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Specification Test Results

Assay (by Metals Basis) 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR