Pearl® Adjustable Micro Pipettes, 12-Channel
Supplier: Heathrow Scientific
Pearl®
120218 120217
77395-224EA 733.53 CAD
77395-224 77395-226
Pearl® Adjustable Micro Pipettes, 12-Channel
Pipettes Micropipettes
Precision liquid handling.
  • Each pipette is calibrated and a calibration certificate is supplied
  • Internal metal parts for increased accuracy and calibration stability
  • Separate tip ejector with the exception of the 500 ml (120105)
  • Large digit volume display
  • Ergonomic, slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • 3-position volume display
  • Volume adjustment and low-force dispensing all in one button for convenience
  • Finger hook
  • Volume locking ring prevents accidental volume changes
  • Color coded and labeled with volume range (both top and side colored components for quick visual confirmation)
  • Precision metal components for both shaft and barrel ensure reliable performance and long life
  • ABS material
The Pearl® is an twelve-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires ²/₃ the operation pressure of most standard pipettes.
Certifications: Manufactured in accordance with ISO 8655 and GMP regulations.
Ordering information: A complete range of single- and multi-channel pipette volumes to cover all your possible needs is available. Contact your Avantor representative for more information.