Aluminium ≥99.999% (metals basis), shot, Puratronic® ∼ 4-8 mm (0.2-0.3 in)
Supplier: Thermo Scientific Chemicals
10573-30
10573-A1
CAAA10573-18EA
176.08
CAD
CAAA10573-18
CAAA10573-30
CAAA10573-A1
Aluminium ≥99.999% (metals basis), shot, Puratronic® ∼ 4-8 mm (0.2-0.3 in)
Aluminium
MDL: MFCD00134029
Purity: 99.999%
CAS No.: 7429-90-5
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Specification Test Results
Assay (by Metals Basis) | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...