Acetic acid ≥99%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Methanecarboxylic acid
, Vinegar acid
, Ethanoic acid
10601-VWNQ09
10601-VWNQ09
10601-VWNQ03
CA87003-241EA
698.14
CAD
87003-241
CA87003-241
87003-239
Acetic acid ≥99%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Acetic acid
High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.
Formula:
H₃CCOOH MW: 60.05 g/mol |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8,II |
Specification Test Results
Assay | ≥ 99 % |
Color | max. 10 APHA |
Aluminum (Al) | max. 1 ppb |
Antimony (Sb) | max. 0.5 ppb |
Arsenic (As) | max. 0.5 ppb |
Barium (Ba) | max. 0.5 ppb |
Beryllium (Be) | max. 0.1 ppb |
Bismuth (Bi) | max. 0.1 ppb |
Cadmium (Cd) | max. 0.5 ppb |
Calcium (Ca) | max. 1 ppb |
Cerium (Ce) | max. 0.1 ppb |
Cesium (Cs) | max. 0.1 ppb |
Chromium (Cr) | max. 1 ppb |
Cobalt (Co) | max. 0.1 ppb |
Copper (Cu) | max. 0.5 ppb |
Dysprosium (Dy) | max. 0.1 ppb |
Erbium (Er) | max. 0.1 ppb |
Europium (Eu) | max. 0.1 ppb |
Gadolinium (Gd) | max. 0.1 ppb |
Gallium (Ga) | max. 0.1 ppb |
Germanium (Ge) | max. 0.5 ppb |
Hafnium (Hf) | max. 0.1 ppb |
Holmium (Ho) | max. 0.1 ppb |
Indium (In) | max. 0.1 ppb |
Iron (Fe) | max. 1 ppb |
Lanthanum (La) | max. 0.1 ppb |
Lead (Pb) | max. 0.1 ppb |
Lithium (Li) | max. 0.1 ppb |
Lutetium (Lu) | max. 0.1 ppb |
Magnesium (Mg) | max. 0.5 ppb |
Manganese (Mn) | max. 0.5 ppb |
Mercury (Hg) | max. 1 ppb |
Molybdenum (Mo) | max. 0.5 ppb |
Neodymium (Nd) | max. 0.1 ppb |
Nickel (Ni) | max. 0.5 ppb |
Platinum (Pt) | max. 0.5 ppb |
Potassium (K) | max. 1 ppb |
Praseodymium (Pr) | max. 0.1 ppb |
Rhenium (Re) | max. 0.1 ppb |
Rhodium (Rh) | max. 0.5 ppb |
Rubidium (Rb) | max. 0.1 ppb |
Ruthenium (Ru) | max. 0.5 ppb |
Samarium (Sm) | max. 0.1 ppb |
Scandium (Sc) | max. 0.1 ppb |
Selenium (Se) | max. 1 ppb |
Silver (Ag) | max. 1 ppb |
Sodium (Na) | max. 1 ppb |
Strontium (Sr) | max. 0.5 ppb |
Tellurium (Te) | max. 0.5 ppb |
Terbium (Tb) | max. 0.1 ppb |
Thallium (Tl) | max. 0.1 ppb |
Thorium (Th) | max. 0.1 ppb |
Thulium (Tm) | max. 0.1 ppb |
Tin (Sn) | max. 0.5 ppb |
Titanium (Ti) | max. 0.5 ppb |
Tungsten (W) | max. 0.5 ppb |
Uranium (U) | max. 0.1 ppb |
Vanadium (V) | max. 0.5 ppb |
Ytterbium (Yb) | max. 0.1 ppb |
Yttrium (Y) | max. 0.1 ppb |
Zinc (Zn) | max. 1 ppb |
Zirconium (Zr) | max. 0.1 ppb |
Chloride (Cl-) | max. 1 ppm |
Phosphate (PO₄³-) | max. 1 ppm |
Substances Reducing Dichromate (K₂Cr₂O₇) | to pass test |
Substances Reducing Permanganate (KMnO₄) | to pass test |
Sulfate (SO₄²-) | max. 0.5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...