SP Bel-Art Sterileware® Sense-able™ Scoops Metal Detectable, Sterile Sampling Tools, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36902-0006 H36904-0006 H36906-0006
89233-982CS 429.62 CAD
89233-982 89233-984 89233-986
SP Bel-Art Sterileware® Sense-able™ Scoops Metal Detectable, Sterile Sampling Tools, Bel-Art Products, a part of SP
Scoops
Why risk your company reputation and suffer costly product recalls? Polystyrene plastic impregnated with stainless steel powder from top to bottom permits scoops and scoop fragments as small as 1.6mm³ to be detected with standard metal detection and X-ray equipment.
  • These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Sterile scoops made with FDA grade materials safe for food, drug, andMore Product Information