VWR® Spatulas with PVC Handles
Supplier: VWR
VWR®
11648-171 11648-177 11648-173 11648-172 11648-175 11648-178 11648-176
82027-516CS 4140.45 CAD
82027-516 82027-506 82027-512 82027-502 82027-514 82027-504 82027-510
VWR® Spatulas with PVC Handles
Spatulas
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • Chemically resistant
  • Durable against boiling water
Spatulas feature stainless steel construction with PVC handles. The handle is resistant to acids and most solvents, and can withstand boiling water. Multiple sizes allow for varied laboratory applications.