Hartmann Mosquito Forceps, Sklar®
Supplier: Sklar
17-1236 17-1336 17-1235 17-1230 17-1238 17-1232 17-1335
82029-966EA 142.13 CAD
82029-966 76192-100 82029-968 76192-194 10194-366 76192-112 10194-326
Hartmann Mosquito Forceps, Sklar®
Forceps
Sklar® Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, they can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Sklar® operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar® instruments meet customer demands. These instruments have been designed to provide exceptional comfort and precision all while ensuring the durability and strength that has been provided by Sklar® for over 100 years.