Tweezers, Reverse Action, Excelta Corp®
Supplier: Excelta
29-SA 30-SA 31-SA 31-SA-C DN-2A-SA-SE DN-3C-SA-SE
63041-854EA 229.97 CAD
63041-854 63041-912 63041-922 63041-928 94024-404 89411-746 89411-748
Tweezers, Reverse Action, Excelta Corp®
Forceps
The reverse action tweezers are closed at rest, allowing for the operator to hold an item without any finger pressure on the tweezer.
  • Will hold parts without finger pressure for extended time
  • Tool determines pressure on component, not operator
Reverse action tweezers are normally closed at rest. This allows the item to be held without hand pressure for any length of time. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Order Now
SPECIFICATIONS
Environmentally Preferable