Littauer Stitch Scissors, Sklar
Supplier: Sklar
15-1355 22-1354
82029-942EA 214.1 CAD
82029-942 89025-248
Littauer Stitch Scissors, Sklar
Scissors
Sklar's® Littauer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Larger, less delicate tip for removing larger sutures
These scissors have a small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer Stitch Scissors have larger, less delicate tips than Spencer Stitch Scissors, and are used for removing larger sutures.
Ordering information: More Product Information