McPherson Vannas Scissors, Sklar
Supplier: Sklar
64-1960 64-1961
82030-294EA 1153.48 CAD
82030-294 82030-296
McPherson Vannas Scissors, Sklar
Scissors
McPherson Vannas Scissors are double spring thumb scissors commonly used in ophthalmic procedures such as iridotomy.
These scissors feature 10 mm, sharp, delicate blades ideal for use on fine tissues of the eye
Censitrac Ready products are industry standard and constructed of stainless steel.
The handle has a horizontally serrated grip that provides greater control of the instrument.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders canMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable