Econo™ Operating Scissors, Floor Grade, Sklar
Supplier: Sklar
21-263 21-266 21-270 21-271 21-272 21-275 21-278 21-281 21-284 21-288 21-291 21-292 21-293 21-294 21-295 21-299 21-300
89023-098PK 100.44 CAD
89023-098 89023-100 89023-106 89023-108 89023-110 89023-112 89023-114 89023-116 89023-118 89023-122 89023-126 89023-130 89023-132 89023-134 89023-136 89023-138 89023-140
Econo™ Operating Scissors, Floor Grade, Sklar
Scissors
Econo™ instruments are floor-grade, disposable instruments.
They eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments reduce the likelihood of cross-contamination.
Made in Pakistan, they offer the great utility.