VWR® Spatulas, PTFE-Coated with Wooden Handles
Supplier: VWR International
82027-472 82027-474 82027-476 82027-478
82027-472CS 3061.75 CAD
82027-472 82027-474 82027-476 82027-478
VWR® Spatulas, PTFE-Coated with Wooden Handles
Spatulas
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • Economical
  • Sturdy construction
  • Non-sticking and non-corrosive
Spatulas feature a PTFE-coated stainless steel blade and a wooden handle. They can be used to transfer sticky or corrosive materials without scratching or damaging container surfaces. Multiple sizes allow for varied laboratory applications.