Iris Tissue Forceps, Sklar
Supplier: Sklar
66-3540 66-3740 66-3340
82030-306EA 179.21 CAD
82030-306 82030-310 82030-308
Iris Tissue Forceps, Sklar
Forceps
Sklar® Iris Tissue Forceps are most commonly used for grasping and handling tissue in ophthalmic procedures.
The narrow tips, however, allow this instrument to be beneficial for use in other microsurgical procedures as well. Iris Tissue Forceps are available with 1 × 2 teeth in multiple lengths and curvatures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.