Metzenbaum Dissecting Scissors, Sklar
Supplier: Sklar
75-5570 75-5670
82030-336EA 218.51 CAD
82030-336 82030-338
Metzenbaum Dissecting Scissors, Sklar
Scissors
Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.