VWR® Hayman Style Spatulas, Stainless Steel
Supplier: VWR International
82027-522 82027-524
82027-522CS 1169.55 CAD
82027-522 82027-524
VWR® Hayman Style Spatulas, Stainless Steel
Spatulas
Stainless steel spatulas ideal for applications involving the transfer or mixing of small sample amounts.
  • Efficient design
  • Stainless steel construction
Model 82027-522 features a round spatula on one end and a spoon on the other. Model 82027-524 features a tapered, bent-tip spatula on one end and a spoon on the other. Efficient design reduces the risk of sample loss.