CentriVap® micro IR Vacuum Concentrator 230 V
Supplier: Labconco
7701030 7701035 7701037
89177-046EA 16558.4 CAD
89177-046 89177-048 89177-054
CentriVap® micro IR Vacuum Concentrator 230 V
Evaporators Centrifugal Evaporators
With the smallest footprint available, the new CentriVap® micro IR Vacuum Concentrator fits in tight spaces and is ideal for small throughput in molecular biology, proteomics, genomics, genetics, cell biology and drug discovery labs
  • 230 V model
Everything about the CentriVap® micro IR is small except its many features. Its transparent infrared (IR) glass lid directs heat to the samples to speed evaporation and reduces cross contamination from condensation. The CentriVap® micro IR includes a rotor for twenty-four small samples, a CentriZap® micro IR strobe light to view samples while the rotor is spinning, an built-in diaphragm pump and glass trap everything you need to start concentrating. Simply plug it in and it's ready to use.
Weighing only 20 pounds, this personal-sized concentrator may be easily transported from lab to lab.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89177-044) is also available, please see separate product page.