Precision Balances, Standard MA-P
Supplier: Mettler Toledo
30697487 30697490 30697493 30697487 30697490 30697493
77323-260EA 2975.42 CAD
77323-260 77323-264 77323-268 CA77323-260 CA77323-264 CA77323-268
Precision Balances, Standard MA-P
Balances and Scales
METTLER TOLEDO Standard MA-P Precision Balances are simple to operate and portable for on-the-go weighing, without compromising on accuracy or performance.
  • Strain gauge weighing cell
  • Internal adjustment
  • LCD hybrid touchscreen
  • 10 built-in applications
  • Power cable and battery operated
  • Front level indicator
  • Overload protection
  • Two interfaces: USB-A, RS232
  • Bluetooth option
The precise weigh cell delivers highly accurate results with each use. Increase efficiency in the workflow with the intuitive touchscreen that guides you through each weighing procedure. Easily export data to a connected printer, computer, or USB device.