Analytical Balances, Advanced MR
Supplier: Mettler Toledo
30666203 30666206 30666209 30666203 30666206 30666209
CA77323-128EA 6660 CAD
CA77323-128 CA77323-132 CA77323-136 77323-128 77323-132 77323-136
Analytical Balances, Advanced MR
Balances and Scales
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
  • Guided leveling
  • Four interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
User management within the 4.5" touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
Order Now
SPECIFICATIONS
Environmentally Preferable
Communication interface USB-A, USB-C, RS232, Ethernet
Voltage 100 - 240 V, 50/60 Hz