Analytical Balances, Advanced MX
Supplier: Mettler Toledo
30665094 30665097 30665140 30665143 30665146 30665094 30665097 30665140 30665143 30665146 30665091
CA77323-040EA 15276.4 CAD
CA77323-040 CA77323-044 CA77323-048 CA77323-052 CA77323-056 77323-040 77323-044 77323-048 77323-052 77323-056 CA77393-646
Analytical Balances, Advanced MX
Balances and Scales
METTLER TOLEDO Advanced MX analytical balances offer a brand new feature- the backlit draft shield. Brightness extends over the draft shield's entire back panel providing exceptional visibility of the work area for highly precise dosing.
  • ErgoDoor
  • User management
  • Leveling assistant
  • StatusLight
  • Three interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The stable SmartPan and high resolution weigh cell and FACT automatic internal adjustment ensure consistently accurate results. Efficient operation within the large, 7" touchscreen includes 8 built-in applications and statistical data analysis. When not using the balance, enable the power-saving mode to reduce energy usage and save costs.