Standard LA Precision Balances
Supplier: Mettler Toledo
30823722 30823723 30853154 30853155 30823722 30823723 30853154 30853155
CA77323-282EA 1870 CAD
CA77323-282 CA77323-284 CA77323-286 CA77323-288 77323-282 77323-284 77323-286 77323-288
Standard LA Precision Balances
Balances and Scales
METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.
  • Front level indicator
  • Overload protection
  • Metal base, ABS top housing
  • RS232 interface
  • Bluetooth option
  • Transfer weight function for easy data transfer
Increase efficiency in daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.