Standard MA Precision Balances
Supplier: Mettler Toledo
30697420 30697423 30697426 30697429 30697440 30697443 30697446 30697449 30697452 30697455 30697458 30697458 30697420 30697423 30697426 30697429 30697440 30697443 30697446 30697449 30697452 30697455
CA77323-200EA 2725 CAD
CA77323-200 CA77323-204 CA77323-208 CA77323-212 CA77323-216 CA77323-220 CA77323-224 CA77323-228 CA77323-232 CA77323-236 CA77323-240 77323-240 77323-200 77323-204 77323-208 77323-212 77323-216 77323-220 77323-224 77323-228 77323-232 77323-236
Standard MA Precision Balances
Balances and Scales
METTLER TOLEDO Standard MA Precision Balances are highly accurate, simple to operate and built with high quality materials for proven long lasting performance.
  • Internal adjustment
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The precise weigh cell and internal adjustment provide accurate results. Intuitive onscreen guidance within the LCD hybrid touchscreen allows you to easily navigate through 10 built in applications and manage weighing data. The metalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Communication interface USB-A, RS232 USB-A, RS232
Voltage 100 - 240 V, 50/60 Hz 100 - 240 V, 50/60 Hz