Ruskin Bone Cutting Rongeurs, Sklar
Supplier: Sklar
40-4512 40-4272 40-4365 40-4372 40-4360
82030-206EA 1929.68 CAD
82030-206 82030-216 82030-212 82030-210 82030-208
Ruskin Bone Cutting Rongeurs, Sklar
Bone Cutters and Rongeurs
The Ruskin Rongeur is a double action, plier handle, debulking instrument used to scoop away bone that may itself be in need of manipulation or may be blocking access to a surgical site.
This is available straight or curved in multiple lengths with multiple bite sizes. Censitrac Ready products are industry standard and constructed of stainless steel. Ruskin Bone Cutting Rongeurs are autoclavable.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.