VITLAB® HDPE Buckets, BrandTech
Supplier: Brandtech
96393 96093 96293 96593
83009-078PK 81.13 CAD
83009-078 83009-076 83009-077 83009-079
VITLAB® HDPE Buckets, BrandTech
Buckets
Buckets feature a hinged handle and graduation markings.
  • HDPE buckets with handles
  • Snap-on PP lids sold separately
  • Durable
These VITLAB® buckets are made of heavy-duty high-density polyethylene for durability and chemical resistance. A hinged handle provides easy gripping and stability. Graduations provide easy estimation of content volumes.
Order Now
SPECIFICATIONS
Environmentally Preferable