SP Bel-Art Sterileware® Red Sterile Sampling Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
H36902-2002 H36906-2008 H36904-2004 H36902-2010 H36904-2010 H36906-2010
47751-004CS 379.19 CAD
47751-004 89140-596 89140-588 47751-008 47751-006 89140-592
SP Bel-Art Sterileware® Red Sterile Sampling Scoops, Bel-Art Products, a part of SP
Scoops
Sterileware® scoops are the perfect shape and size for taking samples, especially from wide-mouth containers.
  • Suited for use with wide-mouth containers
  • Sterile
  • Polystyrene plastic construction
  • Provided in individually sealed polyethylene bag
Designed to hold various capacities, these one-piece sterile scoops are molded from high-impact polystyrene plastic, individually sealed and sterilized in a polyethylene bag, and ready for one-time use and disposal.
The 60 mL (2oz.) and 120 mL (4oz.) sampling scoops have a long handle that increases the distance between the hand and sample. When placed on a flat surface they remain upright and can be used as weighing boats. The bright red color reduces the chance of these scoops being accidentally left behind in a sample.
The distinctive color can also serve as a code to avoid cross contamination and provide a contrasting background to better view light colored samples. Made from FDA-grade materials, they are lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization.