SP Bel-Art Test Tube Baskets, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18738-0000 F18737-0010 F18738-0010 F18739-0010
60914-208CS 414.32 CAD
60914-208 60914-220 60914-263 60914-285 60914-300
SP Bel-Art Test Tube Baskets, Polypropylene, Bel-Art Products, a part of SP
Baskets Baskets for Test Tubes
Ideal for rinsing glassware, these polypropylene baskets are available in three sizes with or without lids.
  • Polypropylene basket construction
  • Easy to drain
  • Autoclavable
  • Available with or without lids
Simply invert lidded baskets for easy draining. Steam autoclavable at 121ºC (250ºF).