CentriVap® Benchtop Centrifugal Concentrators and Systems 230 V
Supplier: Labconco
7810011 7810013 7810015 7812011 7812014 7811030 7460030 7982011 7812012 7812015 7811040 7460040 7982012 7810017 7983014 7983015
89004-918EA 11913.2 CAD
89004-918 89004-922 89004-926 89004-930 89004-934 89004-942 89004-946 89004-950 89004-968 89004-970 89004-972 89004-974 89004-976 89041-238 89041-282 89041-284
CentriVap® Benchtop Centrifugal Concentrators and Systems 230 V
Evaporators Centrifugal Evaporators
Ideal for biology, microbiology, biochemistry, pharmaceutical research, and analytical chemistry laboratories, centrifugal concentrators quickly process the evaporation of multiple small samples.
  • 230 V model
Microprocessor-controlled 300 W heater speeds evaporation by supplying up to 100 °C (212 °F) of heat in 1 °C increments. Optional heat boost warms the chamber with additional heat up to 100 °C in 1 °C increments. Heat and run times are set separately to protect heat-sensitive samples from excessive heat exposure after the solvent has evaporated.More Product Information