VWR® Rochester-Pean Hemostatic Forceps
Supplier: VWR International
82027-418 82027-420 82027-422 82027-424
82027-418CS 3586.34 CAD
82027-418 82027-420 82027-422 82027-424
VWR® Rochester-Pean Hemostatic Forceps
Forceps
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
  • Stainless steel construction
  • Reusable
  • Non-sterile
  • Premium grade
Horizontal serrations help ensure forceps remain clamped tight.