Adson (Ewald) Dressing Forceps, Sklar
Supplier: Sklar
50-3147
82030-278EA 93.04 CAD
82030-278
Adson (Ewald) Dressing Forceps, Sklar
Forceps
Sklar's® Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
Censitrac Ready products are industry standard and constructed of stainless steel.
They have a wide thumb grasp for increased precision and control.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.