InkSmith Climate Action Kits, Energy
Supplier: INKSMITH LIMITED SE
InkSmith
ROB-04-0326 ROB-04-0334
470354-864EA 78.99 CAD
470354-864 470354-860
InkSmith Climate Action Kits, Energy
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Real-world climate investigations, powered by the popular BBC micro:bit!
  • Introduce students to climate change and environmental sustainability topics through engaging problem-based learning activities
  • Students develop technology-related solutions aligned with U.N. Sustainable development goals
  • In-depth curriculum and instructor guides available online
Empower your students to tackle climate change issues while learning important digital skills with the new Energy climate action kit. Just like the popular Land climate action kit, the energy kit will help educators inspire their students to recognize and address climate change issues related to sustainable energy!
Students will explore the United nations sustainable development goals while making connections between them and the climate change issues we're facing. After learning the background and context of each issue, students will design, prototype, and program a solution using the kit components and a BBC micro:bit.
The new Energy climate action kit features three modules: Solar energy, wind energy, and electric vehicles. Each Climate action kit includes a coupon code to access InkSmith's online learning platform filled with tons of classroom-ready resources for educators including interactive Google slide decks and curriculum connections.
Ordering information: Already have micro:bits in your classroom? The Climate action kit, energy is available with or without a micro:bit. The energy add-on pack complements the Climate action kit, land.
Delivery information: Kit includes (1) Climate Action micro:bit Breakout board; (10) alligator clip wire leads; (1) DC motor; (2) wheels; (1) DC fan motor; (1) servo motor; (1) photoresistor sensor; (1) touch sensor; (1) sonar sensor. The kit also includes online access to project-based lessons educator curriculum courses.