VITLAB® PP Buckets, BrandTech
Supplier: Brandtech
96694 96794
83009-080PK 183.39 CAD
83009-080 83009-081
VITLAB® PP Buckets, BrandTech
Buckets
Polypropylene (PP) buckets feature a hinged handle, pouring lip, and graduation markings
  • PP with hinged handle
  • Pouring lip to reduce drips
  • No lids available
Excellent chemical resistance.