SP Bel-Art Tubing Cutter, Plastic, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
Scienceware®
H21010-0000
63022-100CS 811.95 CAD
63022-100
SP Bel-Art Tubing Cutter, Plastic, Bel-Art Products, a part of SP
Cutters
Pocket-sized cutter cuts flexible tubing with an outer diameter up to 19 mm (3/4").
  • Hardened steel blade
  • Jaw is spring-loaded and closes automatically
  • Compact and convenient size
The jaw is lightly spring-loaded and closes automatically for safety. The replaceable, hardened steel blade produces clean, straight cuts.
Ordering information: Replacement blades sold separately.