SP Bel-Art Economy Sampling Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36933-0021 F36933-0024
74950-209PK 37.08 CAD
74950-209 74950-211 74950-213 74950-215 74950-201 74950-203 74950-205 74950-207
SP Bel-Art Economy Sampling Scoops, Bel-Art Products, a part of SP
Scoops
Designed for accurate measurement of small quantities of dry or liquid substances.
  • One-piece design
  • Polystyrene construction
  • Durable
  • Lightweight
  • Flat-bottom shape
One-piece scoops are molded from durable, FDA-grade, high-impact polystyrene plastic. Leveling the sample to the flat top edge provides accurate and repeatable volume measurements. Flat-bottom shape and light weight allow use as a weighing boat.
Order Now
SPECIFICATIONS
Environmentally Preferable