Liston Bone Cutting Forceps, Sklar
Supplier: Sklar
40-3055 40-3077 40-3087 40-3155 40-3177 40-3187
82030-186EA 1221.53 CAD
82030-186 82030-188 82030-190 82030-192 82030-194 10195-628
Liston Bone Cutting Forceps, Sklar
Bone Cutters and Rongeurs
Liston Bone Cutting Forceps are double spring, single action forceps which features with forward or backward angled blades and are commonly used for cutting small bones.
  • Made from premium-grade stainless steel
  • Autoclavable
In order to work with larger bones, use the Liston-Stille Bone Cutting Forceps, which have a similar blade, however, utilize a double action mechanism for greater ability to cut large bones.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.