Littauer Bone Cutting Forceps, Sklar
Supplier: Sklar
97-1251 97-1255 97-1259 97-1286
82030-356EA 336.93 CAD
82030-356 82030-358 82030-360 10194-104
Littauer Bone Cutting Forceps, Sklar
Bone Cutters and Rongeurs
Littauer Cutting Forceps are a non-ratcheted, plier handle instrument used primarily for cutting nail and bone.
  • As nail nippers they are used for nail debridement
  • Product is straight with smooth edges and an overall length of 4 inches
  • Autoclavable and latex-free
They are also be used on delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures
Censitrac Ready products are industry standard and constructed of stainless steel.
The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is particularly helpful with nail debridement which should be completed in short, small sections to avoid damage to the hyponychium.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.